Assay Details
Target Gene Details
Gene Symbol: | lacZ |
Gene Name: | Escherichia coli beta-galactosidase |
Gene Aliases: |
b-gal, beta-gal |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gene Symbol: | lacZ |
Gene Name: | Escherichia coli beta-galactosidase |
Gene Aliases: |
b-gal, beta-gal |