Assay Details
Target Gene Details
Gene Symbol: | cre |
Gene Name: | Enterobacteria phage P1 cyclization recombinase |
Gene Aliases: |
- |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gene Symbol: | cre |
Gene Name: | Enterobacteria phage P1 cyclization recombinase |
Gene Aliases: |
- |