Assay Details
| Gene Symbol: ATHSRP54A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101400.4 | NP_563970.1 | 1 - 2 | 343 | 60 | |
| GenBank mRNA | AF344329.1 | - | 1 - 2 | 232 | 60 | |
| AK221684.1 | - | 1 - 2 | 258 | 60 | ||
| AY086187.1 | - | 1 - 2 | 343 | 60 | ||
More Information
Set Membership: |
|

ATHSRP54A
Genomic Map