Assay Details
| Gene Symbol: AT1G21065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101960.3 | NP_564132.2 | 3 - 4 | 544 | 91 | |
| GenBank mRNA | AF332432.1 | - | 3 - 4 | 449 | 91 | |
| Gene Symbol: EID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK229833.1 | - | 294 | 91 | ||
More Information
Set Membership: |
|

AT1G21065
Genomic Map