Assay Details
| Gene Symbol: AT4G23500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118481.4 | NP_194081.1 | 2 - 3 | 704 | 87 | |
| GenBank mRNA | AK221568.1 | - | 2 - 3 | 576 | 87 | |
| AY142497.1 | - | 2 - 3 | 526 | 87 | ||
More Information
Set Membership: |
|

AT4G23500
Genomic Map