Assay Details
| Gene Symbol: UGT88A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112523.1 | NP_566549.1 | 1 - 2 | 1136 | 86 | |
| NM_112524.4 | NP_566550.1 | 1 - 2 | 1251 | 86 | ||
| NM_180266.3 | NP_850597.1 | 1 - 2 | 1155 | 86 | ||
| GenBank mRNA | AK316752.1 | - | 1 - 2 | 1133 | 86 | |
| AY037255.1 | - | 1 - 2 | 1131 | 86 | ||
| AY143902.1 | - | 1 - 2 | 1115 | 86 | ||
| KJ138822.1 | - | 1 - 2 | 1115 | 86 | ||
More Information
Set Membership: |
|

UGT88A1
Genomic Map