Assay Details
| Gene Symbol: ARC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203006.1 | NP_001189935.1 | 10 - 11 | 1689 | 82 | |
| NM_112862.4 | NP_188606.2 | 10 - 11 | 1658 | 82 | ||
| NM_180284.3 | NP_850615.2 | 10 - 11 | 1658 | 82 | ||
More Information
Set Membership: |
|

ARC5
Genomic Map