Assay Details
| Gene Symbol: ARC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203006.1 | NP_001189935.1 | 2 - 3 | 653 | 77 | |
| NM_112862.4 | NP_188606.2 | 2 - 3 | 730 | 77 | ||
| NM_180284.3 | NP_850615.2 | 2 - 3 | 730 | 77 | ||
More Information
Set Membership: |
|

ARC5
Genomic Map