Assay Details
| Gene Symbol: ARC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203006.1 | NP_001189935.1 | 4 - 5 | 995 | 106 | |
| NM_112862.4 | NP_188606.2 | 4 - 5 | 1072 | 106 | ||
| NM_180284.3 | NP_850615.2 | 4 - 5 | 1072 | 106 | ||
More Information
Set Membership: |
|

ARC5
Genomic Map