Assay Details
| Gene Symbol: CIPK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_099996.4 | NP_171622.1 | 9 - 10 | 1167 | 85 | |
| NM_179239.1 | NP_849570.1 | 9 - 10 | 1024 | 85 | ||
| NM_179240.1 | NP_849571.1 | 9 - 10 | 1018 | 85 | ||
| GenBank mRNA | AF295664.1 | - | 9 - 10 | 911 | 85 | |
| AF339147.1 | - | 9 - 10 | 905 | 85 | ||
More Information
Set Membership: |
|

CIPK9
Genomic Map