Assay Details
Gene Symbol: GIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160826.1 | NP_001154298.1 | 3 - 4 | 488 | 97 | |
NM_099998.4 | NP_563619.1 | 2 - 3 | 404 | 97 | ||
GenBank mRNA | AF410307.1 | - | 2 - 3 | 324 | 97 | |
AY102133.1 | - | 2 - 3 | 160 | 97 | ||
AY102640.1 | - | 2 - 3 | 364 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
