Assay Details
| Gene Symbol: SLP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335625.1 | NP_001324171.1 | 8 - 9 | 1672 | 90 | |
| NM_127474.4 | NP_565447.1 | 8 - 9 | 1794 | 90 | ||
| GenBank mRNA | AY051009.1 | - | 8 - 9 | 1820 | 90 | |
| AY133826.1 | - | 7 - 8 | 1485 | 90 | ||
More Information
Set Membership: |
|

SLP3
Genomic Map