Assay Details
Gene Symbol: SYP31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120658.5 | NP_196195.1 | 3 - 4 | 800 | 107 | |
GenBank mRNA | AY054184.1 | - | 3 - 4 | 731 | 107 | |
AY066041.1 | - | 3 - 4 | 613 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
