Assay Details
| Gene Symbol: SYP31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120658.5 | NP_196195.1 | 5 - 6 | 1031 | 88 | |
| GenBank mRNA | AY054184.1 | - | 5 - 6 | 962 | 88 | |
| AY066041.1 | - | 5 - 6 | 844 | 88 | ||
More Information
Set Membership: |
|

SYP31
Genomic Map