Assay Details
| Gene Symbol: BBX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127704.3 | NP_565507.1 | 4 - 5 | 707 | 75 | |
| GenBank mRNA | AY060570.1 | - | 4 - 5 | 637 | 75 | |
| AY125573.1 | - | 4 - 5 | 468 | 75 | ||
More Information
Set Membership: |
|

BBX18
Genomic Map