Assay Details
Gene Symbol: ATPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035768.1 | NP_001030845.1 | 1 - 2 | 293 | 96 | |
NM_115090.4 | NP_190798.1 | 1 - 2 | 434 | 96 | ||
GenBank mRNA | AY050330.1 | - | 1 - 2 | 244 | 96 | |
AY086788.1 | - | 1 - 2 | 293 | 96 | ||
AY094036.1 | - | 1 - 2 | 169 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
