Assay Details
| Gene Symbol: ATPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035768.1 | NP_001030845.1 | 4 - 5 | 533 | 75 | |
| NM_115090.4 | NP_190798.1 | 4 - 5 | 670 | 75 | ||
| GenBank mRNA | AY050330.1 | - | 4 - 5 | 480 | 75 | |
| AY086788.1 | - | 4 - 5 | 529 | 75 | ||
| AY094036.1 | - | 4 - 5 | 405 | 75 | ||
More Information
Set Membership: |
|

ATPQ
Genomic Map