Assay Details
Gene Symbol: AGK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035800.1 | NP_001030877.1 | 6 - 7 | 1020 | 75 | |
NM_001339877.1 | NP_001325466.1 | 6 - 7 | 837 | 75 | ||
NM_115615.3 | NP_567051.1 | 6 - 7 | 1134 | 75 | ||
GenBank mRNA | AF204676.1 | - | 6 - 7 | 1020 | 75 | |
BT002466.1 | - | 6 - 7 | 931 | 75 | ||
BT009693.1 | - | 4 - 5 | 687 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
