Assay Details
| Gene Symbol: THO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123657.3 | NP_568616.1 | 3 - 4 | 1329 | 110 | |
| NM_203144.4 | NP_974873.1 | 4 - 5 | 1345 | 110 | ||
| GenBank mRNA | AY039925.1 | - | 3 - 4 | 1295 | 110 | |
| AY091346.1 | - | 3 - 4 | 866 | 110 | ||
More Information
Set Membership: |
|

THO5
Genomic Map