Assay Details
| Gene Symbol: LAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339986.1 | NP_001325620.1 | 1 - 2 | 602 | 65 | |
| NM_115815.6 | NP_191512.1 | 2 - 3 | 530 | 65 | ||
| NM_202733.3 | NP_974462.1 | 2 - 3 | 466 | 65 | ||
More Information
Set Membership: |
|

LAP3
Genomic Map