Assay Details
| Gene Symbol: AT5G12980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121301.5 | NP_196802.1 | 6 - 7 | 667 | 72 | |
| GenBank mRNA | AY161167.1 | - | 6 - 7 | 611 | 72 | |
More Information
Set Membership: |
|

AT5G12980
Genomic Map