Assay Details
| Gene Symbol: KT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336238.1 | NP_001324058.1 | 3 - 4 | 1075 | 93 | |
| NM_128562.4 | NP_180568.1 | 3 - 4 | 770 | 93 | ||
| GenBank mRNA | AF029876.1 | - | 3 - 4 | 541 | 93 | |
| AF033118.1 | - | 3 - 4 | 575 | 93 | ||
| AK221387.1 | - | 3 - 4 | 637 | 93 | ||
More Information
Set Membership: |
|

KT1
Genomic Map