Assay Details
| Gene Symbol: KT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336238.1 | NP_001324058.1 | 7 - 8 | 1558 | 106 | |
| NM_128562.4 | NP_180568.1 | 7 - 8 | 1253 | 106 | ||
| GenBank mRNA | AF029876.1 | - | 7 - 8 | 1024 | 106 | |
| AF033118.1 | - | 7 - 8 | 1058 | 106 | ||
| AK221387.1 | - | 7 - 8 | 1120 | 106 | ||
More Information
Set Membership: |
|

KT1
Genomic Map