Assay Details
Gene Symbol: SAG29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121320.3 | NP_196821.1 | 1 - 2 | 233 | 74 | |
GenBank mRNA | AY045949.1 | - | 1 - 2 | 186 | 74 | |
AY113934.1 | - | 1 - 2 | 50 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
