Assay Details
| Gene Symbol: DRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121466.3 | NP_196966.2 | 6 - 7 | 679 | 114 | |
| GenBank mRNA | AF240695.1 | - | 6 - 7 | 632 | 114 | |
| AK176138.1 | - | 6 - 7 | 674 | 114 | ||
| AK220953.1 | - | 6 - 7 | 674 | 114 | ||
| AK229197.1 | - | 6 - 7 | 679 | 114 | ||
More Information
Set Membership: |
|

DRM2
Genomic Map