Assay Details
| Gene Symbol: GNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121582.2 | NP_197081.1 | 1 | 371 | 119 | |
| Gene Symbol: PSRP3/2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121581.3 | NP_197080.1 | 2 | 967 | 119 | |
More Information
Set Membership: |
|

GNA1
Genomic Map