Assay Details
Gene Symbol: SOBIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128746.3 | NP_180747.1 | 1 | 1274 | 122 | |
GenBank mRNA | AF370596.1 | - | 1 | 1248 | 122 | |
FJ708707.1 | - | 1 | 1248 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
