Assay Details
| Gene Symbol: PPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116399.4 | NP_192078.1 | 2 - 3 | 625 | 63 | |
| NM_178952.2 | NP_849283.1 | 2 - 3 | 601 | 63 | ||
| GenBank mRNA | BT020418.1 | - | 2 - 3 | 559 | 63 | |
| BT020535.1 | - | 2 - 3 | 578 | 63 | ||
| D83139.1 | - | 2 - 3 | 559 | 63 | ||
More Information
Set Membership: |
|

PPOX
Genomic Map