Assay Details
| Gene Symbol: EMB2369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116673.7 | NP_192344.1 | 17 - 18 | 2510 | 79 | |
| GenBank mRNA | AY079159.1 | - | 17 - 18 | 2510 | 79 | |
| BT002249.1 | - | 17 - 18 | 2471 | 79 | ||
More Information
Set Membership: |
|

EMB2369
Genomic Map