Assay Details
| Gene Symbol: CRK38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340486.1 | NP_001328736.1 | 3 - 4 | 1108 | 99 | |
| NM_116689.2 | NP_192360.1 | 3 - 4 | 1058 | 99 | ||
More Information
Set Membership: |
|

CRK38
Genomic Map