Assay Details
| Gene Symbol: AMT1;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117425.2 | NP_193087.1 | 1 | 1539 | 175 | |
| GenBank mRNA | X75879.1 | - | 1522 | 176 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AMT1;1| Gene Symbol: AMT1;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117425.2 | NP_193087.1 | 1 | 1539 | 175 | |
| GenBank mRNA | X75879.1 | - | 1522 | 176 | ||
Set Membership: |
|