Assay Details
Gene Symbol: GR-RBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084916.1 | NP_001078385.1 | 3 - 4 | 258 | 81 | |
NM_001084917.1 | NP_001078386.1 | 3 - 4 | 258 | 81 | ||
NM_117459.4 | NP_193121.1 | 3 - 4 | 496 | 81 | ||
NM_179046.2 | NP_849377.1 | 3 - 4 | 258 | 81 | ||
GenBank mRNA | AY072361.1 | - | 3 - 4 | 254 | 81 | |
AY085621.1 | - | 3 - 4 | 257 | 81 | ||
BT002197.1 | - | 2 - 3 | 188 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
