Assay Details
| Gene Symbol: ABCG30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341010.1 | NP_001319944.1 | 12 - 13 | 2564 | 83 | |
| NM_001341011.1 | NP_001328800.1 | 12 - 13 | 2570 | 83 | ||
| NM_001341012.1 | NP_001328801.1 | 13 - 14 | 2270 | 83 | ||
| NM_117611.5 | NP_193258.3 | 12 - 13 | 2570 | 83 | ||
More Information
Set Membership: |
|

ABCG30
Genomic Map