Assay Details
| Gene Symbol: DIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334269.1 | NP_001319330.1 | 3 - 4 | 730 | 78 | |
| NM_001334270.1 | NP_001321710.1 | 2 - 3 | 712 | 78 | ||
| NM_105377.5 | NP_176878.1 | 2 - 3 | 1003 | 78 | ||
| GenBank mRNA | AK176481.1 | - | 2 - 3 | 566 | 78 | |
| BT025995.1 | - | 2 - 3 | 523 | 78 | ||
More Information
Set Membership: |
|

DIN9
Genomic Map