Assay Details
Gene Symbol: NF-YC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123174.5 | NP_198630.2 | 2 - 3 | 498 | 80 | |
GenBank mRNA | BT021919.1 | - | 2 - 3 | 246 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
