Assay Details
Gene Symbol: NF-YC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123174.5 | NP_198630.2 | 5 - 6 | 849 | 78 | |
GenBank mRNA | BT021919.1 | - | 5 - 6 | 597 | 78 | |
BT025775.1 | - | 4 - 5 | 464 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
