Assay Details
| Gene Symbol: SERK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126955.5 | NP_178999.2 | 2 - 3 | 622 | 125 | |
| GenBank mRNA | AY056243.1 | - | 2 - 3 | 262 | 125 | |
| FJ708691.1 | - | 2 - 3 | 235 | 125 | ||
More Information
Set Membership: |
|

SERK4
Genomic Map