Assay Details
| Gene Symbol: SERK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126955.5 | NP_178999.2 | 6 - 7 | 982 | 90 | |
| GenBank mRNA | AY056243.1 | - | 6 - 7 | 622 | 90 | |
| FJ708691.1 | - | 6 - 7 | 595 | 90 | ||
More Information
Set Membership: |
|

SERK4
Genomic Map