Assay Details
| Gene Symbol: DAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111334.6 | NP_187113.1 | 2 - 3 | 500 | 88 | |
| GenBank mRNA | BT010655.1 | - | 2 - 3 | 276 | 88 | |
| BT010971.1 | - | 2 - 3 | 222 | 88 | ||
More Information
Set Membership: |
|

DAN1
Genomic Map