Assay Details
| Gene Symbol: ADT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337726.1 | NP_001325505.1 | 10 - 11 | 1118 | 73 | |
| NM_111642.3 | NP_187420.1 | 10 - 11 | 1136 | 73 | ||
| NM_202520.2 | NP_974249.1 | 10 - 11 | 1099 | 73 | ||
| GenBank mRNA | AY050813.1 | - | 10 - 11 | 1094 | 73 | |
| AY113967.1 | - | 10 - 11 | 1087 | 73 | ||
| DQ411465.1 | - | 10 - 11 | 1091 | 73 | ||
More Information
Set Membership: |
|

ADT2
Genomic Map