Assay Details
Gene Symbol: NF-YC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112083.4 | NP_187854.2 | 3 - 4 | 503 | 94 | |
GenBank mRNA | AF462841.1 | - | 3 - 4 | 429 | 94 | |
AY143847.1 | - | 3 - 4 | 201 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
