Assay Details
Gene Symbol: EXT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106344.3 | NP_565143.1 | 2 - 3 | 1043 | 115 | |
NM_179564.1 | NP_849895.1 | 3 - 4 | 760 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
