Assay Details
| Gene Symbol: BGLU17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337088.1 | NP_001324585.1 | 2 - 3 | 345 | 72 | |
| NM_001337089.1 | NP_001324587.1 | 2 - 3 | 345 | 72 | ||
| NM_130011.3 | NP_181976.1 | 2 - 3 | 284 | 72 | ||
| GenBank mRNA | AY074629.1 | - | 2 - 3 | 255 | 72 | |
More Information
Set Membership: |
|

BGLU17
Genomic Map