Assay Details
| Gene Symbol: BGLU17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125053.1 | NP_001118525.1 | 4 - 5 | 703 | 105 | |
| NM_001337088.1 | NP_001324585.1 | 5 - 6 | 568 | 105 | ||
| NM_001337089.1 | NP_001324587.1 | 5 - 6 | 568 | 105 | ||
| NM_001337090.1 | NP_001324586.1 | 1 - 2 | 105 | 105 | ||
| NM_130011.3 | NP_181976.1 | 5 - 6 | 507 | 105 | ||
| GenBank mRNA | AY074629.1 | - | 5 - 6 | 478 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001337090.1 transcripts.
Set Membership: |
|

BGLU17
Genomic Map