Assay Details
| Gene Symbol: BGLU17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125053.1 | NP_001118525.1 | 8 - 9 | 1368 | 79 | |
| NM_001337088.1 | NP_001324585.1 | 9 - 10 | 1233 | 79 | ||
| NM_001337090.1 | NP_001324586.1 | 4 - 5 | 847 | 79 | ||
| NM_130011.3 | NP_181976.1 | 9 - 10 | 1172 | 79 | ||
| GenBank mRNA | AY074629.1 | - | 9 - 10 | 1143 | 79 | |
More Information
Set Membership: |
|

BGLU17
Genomic Map