Assay Details
| Gene Symbol: LTP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125323.5 | NP_568905.1 | 1 - 2 | 434 | 79 | |
| GenBank mRNA | AY065293.1 | - | 1 - 2 | 414 | 79 | |
| AY094428.1 | - | 1 - 2 | 416 | 79 | ||
| AY096572.1 | - | 1 - 2 | 337 | 79 | ||
More Information
Set Membership: |
|

LTP3
Genomic Map