Assay Details
| Gene Symbol: ALATS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198257.1 | NP_001185186.2 | 13 - 14 | 1917 | 112 | |
| NM_103905.4 | NP_175439.2 | 13 - 14 | 2007 | 112 | ||
More Information
Set Membership: |
|

ALATS
Genomic Map