Assay Details
| Gene Symbol: CYS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121252.3 | NP_196775.1 | 2 - 3 | 453 | 79 | |
| GenBank mRNA | AY088332.1 | - | 2 - 3 | 418 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYS1| Gene Symbol: CYS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121252.3 | NP_196775.1 | 2 - 3 | 453 | 79 | |
| GenBank mRNA | AY088332.1 | - | 2 - 3 | 418 | 79 | |
Set Membership: |
|