Assay Details
| Gene Symbol: KT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336078.1 | NP_001324538.1 | 9 - 10 | 1782 | 55 | |
| NM_128222.6 | NP_180233.1 | 10 - 11 | 2037 | 55 | ||
| GenBank mRNA | AK317729.1 | - | 10 - 11 | 1808 | 55 | |
| BT006442.1 | - | 9 - 10 | 1428 | 55 | ||
| X62907.1 | - | 1800 | 55 | |||
More Information
Set Membership: |
|

KT1
Genomic Map