Assay Details
| Gene Symbol: KT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336078.1 | NP_001324538.1 | 1 - 2 | 564 | 67 | |
| NM_128222.6 | NP_180233.1 | 2 - 3 | 819 | 67 | ||
| GenBank mRNA | AK317729.1 | - | 2 - 3 | 590 | 67 | |
| BT006442.1 | - | 1 - 2 | 210 | 67 | ||
| X62907.1 | - | 582 | 67 | |||
More Information
Set Membership: |
|

KT1
Genomic Map